Anti-CCR5 Rat Monoclonal Antibody [clone: HEK/1/85a]
Catalog # USBIC2099-99K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) receptor 5(gene/pseudogene)
- Antigen symbol:CCR5
- Clonality:Monoclonal
- Clone:HEK/1/85a
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:50-1:100
Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: Monoclonal
Clone: HEK/1/85a
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human