Anti-BRE Rabbit polyclonal antibody unconjugated
USBIB0085-04E
- Antibody type:Primary
- Antigen name:brain and reproductive organ-expressed (TNFRSF1A modulator)
- Antigen symbol:BRE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: BRE (brain and reproductive organ-expressed (TNFRSF1A modulator))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human