Anti-MEX3A Rabbit Polyclonal Antibody
USBI146333
- Antibody type:Primary
- Antigen name:Mex-3 RNA binding family member A
- Antigen symbol:MEX3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1 ?g/ml; Immunofluorescence: 20 ?g/ml; Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: MEX3A (mex-3 RNA binding family member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat