United Kingdom
Order Entry
Anti-CD2 Mouse Monoclonal Antibody [clone: 13H76]
Anti-CD2 Mouse Monoclonal Antibody [clone: 13H76]
  USBI137662
Anti-CD2 Mouse Monoclonal Antibody [clone: 13H76]
  USBI137662
 
£664.00
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    CD2 molecule
  • Antigen symbol:
    CD2
  • Clonality:
    Monoclonal
  • Clone:
    13H76
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Format:
    ascites
  • Form:
    liquid
  • Storage buffer:
    20mMTRIS, 150mM sodium chloride, 0,001% merthiolate.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: CD2 (CD2 molecule)
Clonality: monoclonal
Clone: 13H76
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human