Anti-WDR34 Mouse Polyclonal Antibody
USBI135345
- Antibody type:Primary
- Antigen name:WD repeat domain 34
- Antigen symbol:WDR34
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 419
- Storage buffer:PBS, pH 7,2.
- Sequence:MVIRELNKNWQSHAFDGFEVNWTEQQQMVSCLYTLGYPPAQAQGLHVTSISWNSTGSVVACAYGRLDHGDWSTLKSFVCAWNLDRRDLRPQQPSAVVEVPSAVLCLAFHPTQPSHVAGGLYSGEVLVWDLSRLEDPLLWRTGLTDDTHTDPVSQVVWLPEPGHSHRFQVLSVATDGKVLLWQGIGVGQLQLTEGFALVMQQLPRSTKLKKHPRGETEVGATAVAFSSFDPRLFILGTEGGFPLKCSLAAGEAALTRMPSSVPLRAPAQFTFSPHGGPIYSVSCSPFHRNLFLSAGTDGHVHLYSMLQAPPLTSLQLSLKYLFAVRWSPVRPLVFAAASGKGDVQLFDLQKSSQKPTVLIKQTQDESPVYCLEFNSQQTQLLAAGDAQGTVKVWQLSTEFTEQGPREAEDLDCLAAEVAA
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: WDR34 (WD repeat domain 34)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human