Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPAG5 Mouse Monoclonal Antibody [clone: 5F9]

Supplier: US Biological
Type: Primary
Antigen: SPAG5 (sperm associated antigen 5)
Clonality: monoclonal
Clone: 5F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG USBI133742
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol SPAG5
Antigen name sperm associated antigen 5
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 5F9
Host Mouse
Isotype IgG3 kappa
Western blot Yes
ELISA Yes
Purification purified by Protein A chromatography
Sequence AETETKVLQEALAGQLDSNCQPMATNWIQEKVWLSQEVDKLRVMFLEMKNEKEKLMIKFQSHRNILEENLRRSDKELEKLDDIVQHIYKTLLSIPEVVRGCKELQGLLEF
Form liquid
Storage buffer PBS, pH 7,2.
Storage temperature Cool
Shipping temperature Ice

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty