Anti-SOCS6 Mouse Monoclonal Antibody [clone: M2-F12]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI133692EA 490 GBP
USBI133692
Anti-SOCS6 Mouse Monoclonal Antibody [clone: M2-F12]
Antibodies
Type: Primary
Antigen: SOCS6 (suppressor of cytokine signaling 6)
Clonality: monoclonal
Clone: M2-F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR