Anti-SOCS5 Mouse Monoclonal Antibody [clone: 2D1]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI133690EA 490 GBP
USBI133690
Anti-SOCS5 Mouse Monoclonal Antibody [clone: 2D1]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: SOCS5 (suppressor of cytokine signaling 5)
Clonality: monoclonal
Clone: 2D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR