Anti-SLAMF7 Mouse Monoclonal Antibody [clone: 1B9]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI133392EA 490 GBP
USBI133392
Anti-SLAMF7 Mouse Monoclonal Antibody [clone: 1B9]
Antibodies
Type: Primary
Antigen: SLAMF7 (SLAM family member 7)
Clonality: monoclonal
Clone: 1B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR