Anti-FGL2 Rabbit Polyclonal Antibody
USBI126801
- Antibody type:Primary
- Antigen name:Fibrinogen-like 2
- Antigen symbol:FGL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 439
- Storage buffer:PBS, pH 7,2.
- Sequence:MKLANWYWLSSAVLATYGFLVVANNETEEIKDERAKDVCPVRLESRGKCEEAGECPYQVSLPPLTIQLPKQFSRIEEVFKEVQNLKEIVNSLKKSCQDCKLQADDNGDPGRNGLLLPSTGAPGEVGDNRVRELESEVNKLSSELKNAKEEINVLHGRLEKLNLVNMNNIENYVDSKVANLTFVVNSLDGKCSKCPSQEQIQSRPVQHLIYKDCSDYYAIGKRSSETYRVTPDPKNSSFEVYCDMMGGGWTVLQARLDGSTNFTRTWQDYKAGFGNLRREFWLGNDKIHLLTKSKEMILRIDLEDFNGVELYALYDQFYVANEFLKYRLHVGNYNGTAGDALRFNKHYNHDLKFFTTPDKDNDRYPSGNCGLYYSSGWWFDACLSANLNGKYYHQKYRGVRNGIFWGTWPGVSEAHPGGYKSSFKEAKMMIRPKHFKP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: FGL2 (fibrinogen-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human