United Kingdom
Order Entry
Anti-DCLK1 Rabbit Polyclonal Antibody
Catalog # USBI034506
Anti-DCLK1 Rabbit Polyclonal Antibody
  USBI034506
 
£626.00
   
  1 * 200 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    doublecortin-like kinase 1
  • Antigen symbol:
    DCLK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Form:
    liquid
  • Amino acid number:
    152 to 181
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: DCLK1 (doublecortin-like kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse