Anti-BIRC3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
USBI032524-APC
- Antibody type:Primary
- Antigen name:baculoviral IAP repeat containing 3
- Antigen symbol:BIRC3
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Amino acid number:58 to 88
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: BIRC3 (baculoviral IAP repeat containing 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human