Anti-MAP2 Mouse Monoclonal Antibody [clone: 13B492]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI030414EA 554 GBP
USBI030414
Anti-MAP2 Mouse Monoclonal Antibody [clone: 13B492]
Antibodies
Type: Primary
Antigen: MAP2 (microtubule-associated protein 2)
Clonality: Monoclonal
Clone: 13B492
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR