Anti-Lipoma Preferred Partner, LPP Mouse Monoclonal Antibody [clone: 13B484]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI030406EA 532 GBP
USBI030406
Anti-Lipoma Preferred Partner, LPP Mouse Monoclonal Antibody [clone: 13B484]
Antibodies
Type: Primary
Antigen: Lipoma Preferred Partner, LPP
Clonality: Monoclonal
Clone: 13B484
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR