Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KSHV ORF62 Mouse Monoclonal Antibody [clone: 13B468]

Supplier: US Biological
Type: Primary
Antigen: KSHV ORF62
Clonality: Monoclonal
Clone: 13B468
Conjugation: Unconjugated
Host: Mouse
Isotype: IgM

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µl 100 µl USBI030390
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol KSHV ORF62
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 13B468
Host Mouse
Isotype IgM
Western blot Yes
Format ascites
Storage buffer 0,03% sodium azide.
Storage temperature Cool
Shipping temperature Ice


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web. will publish your name as user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty