Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Mouse IgG (H+L) Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
Anti-Mouse IgG (H&L) biotin conjugated antibody is suitable for highly specific immunological methods requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:350000; Western Blot: 1:2000 - 1:20000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
2 mg 2 mg ROCK610-1602
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol IgG (H+L)
Antigen name IgG (H+L)
Reactivity Mouse
Conjugation Biotin
Clonality Polyclonal
Antibody type Secondary
Host Goat
Isotype none
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Gel shift Yes
Flow cytometry Yes
ChIP Yes
Neutralisation Yes
Concentration 2,0 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross adsorption non
Immunogen Anti-Mouse IgG was produced by repeated immunization with Mouse IgG whole molecule in goat.
Purification purified by immunoaffinity chromatography
Form Lyophilized
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Storage temperature Cool
Shipping temperature Ice

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty