|
|
ORDER
SPECIFICATIONS
Antigen symbol | IgM µ |
Antigen name | IgM µ |
Reactivity | Mouse |
Conjugation | Peroxidase |
Clonality | Polyclonal |
Antibody type | Secondary |
Host | Goat |
Isotype | none |
Western blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
ImmunoPrecipitation | Yes |
Gel shift | Yes |
Flow cytometry | Yes |
ChIP | Yes |
Neutralisation | Yes |
Concentration | 1,0 mg/ml by UV absorbance at 280 nm |
Format | Antibody fraction |
Cross adsorption | non |
Immunogen | Anti-Mouse IgM (mu chain) was produced by repeated immunization with mouse IgM mu heavy chain in goat. |
Purification | purified by immunoaffinity chromatography |
Form | Lyophilized |
Storage buffer | 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free |
Storage temperature | Cool |
Shipping temperature | Ice |
WRITE A REVIEW
REVIEWS (0)
Sort all by: