Anti-LEP Rabbit Polyclonal Antibody
Catalog # ROCK110-401-320
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:leptin
- Antigen symbol:LEP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:16846
- Antigen synonyms:obese|ob
- Storage buffer:0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:This whole rabbit serum was prepared by repeated immunizations with recombinant mouse leptin 16000 MW produced in E. coli.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
This antiserum against Mouse Leptin has been tested for use in ELISA and immunoblotting. Reactivity in other immunoassays is unknown. Obesity associated leptin is the product of the OB gene and has been identified with Type II diabetes.
Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Human