Anti-GAPDH Goat polyclonal antibody unconjugated

Supplier: GenScript Biotech
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

GENSA00191-40EA 153 GBP
GENSA00191-40
Anti-GAPDH Goat polyclonal antibody unconjugated
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR