Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Restin/CLIP1 Goat Polyclonal Antibody

Supplier: Thermo Scientific

Type: Primary
Antigen: Restin/CLIP1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG PIEAPA519167
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CLIP1
Antigen name CAP-GLY Domain containing linker Protein 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Goat
Isotype none
Western Blot Yes
ImmunoChemistry Yes
Antigen synonyms CLIP-170, CAP-GLY domain containing linker protein 1, CYLN1, CLIP, CLIP170, CLIP1, MGC131604, restin, RSN, Reed-Steinberg cell-expressed intermediate filament-associated protein, cytoplasmic linker 1, restin (Reed-Steinberg cell-expressed intermediate filament-associated protein)
Immunogen Synthetic peptide
Storage Buffer TBS, pH 7,3, with 0,5% BSA and 0,02% Sodium azide
Storage temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty