Anti-Serotonin Receptor 3B/HTR3B Goat Polyclonal Antibody

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PIEAPA519163EA 0 GBP
PIEAPA519163
Anti-Serotonin Receptor 3B/HTR3B Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Serotonin Receptor 3B/HTR3B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR