Anti-TUBB Rabbit Polyclonal Antibody

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PIEAPA116947EA 0 GBP
PIEAPA116947
Anti-TUBB Rabbit Polyclonal Antibody
Antibodies
Suggested positive control: antigen standard for TUBB (transient overexpression lysate), Hela whole cell extract. Tubulin is the major building block of microtubules. This intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules function as structural and mobile elements in mitosis, intracellular transport, flagellar movement, and in the cytoskeleton. Antibodies against beta Tubulin are useful as loading controls for Western Blotting.

Type: Primary
Antigen: TUBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Primates, Hamster, Human, Chicken, Bovine, Zebrafish, Xenopus laevis, Rat, Porcine, Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR