Anti-Calreticulin Rabbit Polyclonal Antibody

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PIEAPA116924EA 0 GBP
PIEAPA116924
Anti-Calreticulin Rabbit Polyclonal Antibody
Antibodies
PA1-16924 is specific for the ~55 kDa Calreticulin protein. There are also several reported isoforms that range from 48-62 kDa. Suggested positive control: antigen standard for CALR (transient overexpression lysate). Calreticulin is a multifunctional, highly conserved Ca2+ -binding protein that is localized to the endoplasmic reticulum (ER). It is also a molecular chaperone and is involved in protein folding events in the ER. Calreticulin has also been shown to interact with the cytoskeleton and to be involved in the regulation of gene expression. Calreticulin may play a role in cellular proliferation including its apparent activity in the proliferation of certain viruses within mammalian host cells. Recently it has been shown that calreticulin is induced in response to various types of cell stress including amino acid deprivation. Close interconnections among protein synthesis, gene expression and calcium signaling have been observed by many researchers in recent years. Calreticulin might be centrally located and crucially participate in the coordination of these functions by the cell.

Type: Primary
Antigen: Calreticulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR