Anti-JARID2 Rabbit Polyclonal Antibody

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PIEAPA116916EA 0 GBP
PIEAPA116916
Anti-JARID2 Rabbit Polyclonal Antibody
Antibodies
PA1-16916 is specific for the ~140 kDa Jumonji protein. Another band is seen >200 kDa and a faint band may be seen ~80 kDa. Jumonji is a nuclear protein that participates in the negative regulation of cell growth. It is required for neural tube formation and is essential for normal heart development and function. As well, it is a part of a group of proteins characterized by a novel structural motif, the JmjC domain, which is implicated in chromatin regulation through histone demethylation.

Type: Primary
Antigen: JARID2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR