The nuclear receptor coactivators participate in the transcriptional activation of specific genes by nuclear receptors. Using a coactivator peroxisome proliferator-activated receptor-interacting protein (PRIP) as bait, a novel, protein has been identified, designated PRIP-interacting protein with methyltransferase domain (PIMT). It is expressed in heart, skeletal muscle, kidney, liver, and placenta and is a component of nuclear receptor signal transduction apparatus acting through PRIP.
Type: Primary
Antigen: PIMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human