Anti-CD36 Rabbit Polyclonal Antibody

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PIEAPA116813EA 0 GBP
PIEAPA116813
Anti-CD36 Rabbit Polyclonal Antibody
Antibodies
This antibody shares 95% sequence homology with rat. CD36 is a cell surface class B scavenger receptor. CD36 is preferentially found within lipid rafts, which facilitates its association with receptors, signalling and adapter molecules. CD36 binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. May function as a cell adhesion molecule. Directly mediates cyto adherence of Plasmodium falciparum parasitized erythrocytes. Binds long chain fatty acids and may function in the transport and/or as a regulator of fatty acid transport.

Type: Primary
Antigen: CD36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Bovine, Primates, Porcine, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR