Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC7A11 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific

Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µl 100 µl PIEAPA116775
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol Solute carrier family 7, member 11
Antigen name Solute Carrier Family 7 Member 11
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype none
Western Blot Yes
Antigen synonyms Amino Acid Transport System Xc-, Solute Carrier Family 7 (Anionic Amino Acid Transporter Light Chain, Member 11, Xc- System), CCBR1,Calcium Channel Blocker Resistance Protein
Storage Buffer Tris glycine with 0,15M NaCl and 0,05% Sodium azide
Storage temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty