Anti-Dishevelled 2 Rabbit Polyclonal Antibody
ABCAAB137528-50
: Abcam
- Antibody type:Primary
- Antigen name:Dishevelled 2
- Antigen symbol:DVL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl