Anti-MPP5 Rabbit Polyclonal Antibody

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABCAAB106850-100EA 613 GBP
ABCAAB106850-100
Anti-MPP5 Rabbit Polyclonal Antibody
Type: Primary
Antigen: MPP5 (membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR