Anti-BAZ1B Rabbit polyclonal antibody

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABCAAB50850-100EA 595 GBP
ABCAAB50850-100
Anti-BAZ1B Rabbit polyclonal antibody
Type: Primary
Antigen: BAZ1B (bromodomain adjacent to zinc finger domain, 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR