Anti-MYO9B Goat Polyclonal Antibody
BIRBORB19278-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Myosin 9B
- Antigen symbol:MYO9B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA._x000D_
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide (Internal Region)
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: MYO9B (myosin IXB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human