Anti-HEXIM1 Mouse Monoclonal Antibody [clone: 1H5]

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABNOH00010614-M10EA 390 GBP
ABNOH00010614-M10
Anti-HEXIM1 Mouse Monoclonal Antibody [clone: 1H5]
Antibodies
Type: Primary
Antigen: HEXIM1
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR