Anti-AGPAT2 Mouse Polyclonal Antibody

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABNOH00010555-A01EA 255 GBP
ABNOH00010555-A01
Anti-AGPAT2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AGPAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR