Anti-IPO8 Rabbit Polyclonal Antibody
AVIVARP65771_P050
- Antibody type:Primary
- Antigen name:importin 8
- Antigen symbol:IPO8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10526
- Antigen synonyms:RANBP8
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: IPO8 (importin 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human