PerfectBlue™ Gel System Maxi M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PEQL41-2025EA 821 GBP
PEQL41-2025
PerfectBlue™ Gel System Maxi M
Wider than the Maxi S Plus, the Maxi M model can accommodate up to 168 samples but still in an economical footprint.
*Based on an agarose gel of approximately 5 mm thickness
Ordering information: Supplied with three combs (1,5 mm thickness, 16, 24, 36 teeth), lid, gel tray, three point levelling platform, 'end gates', tank and cables.