Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cleanroom tables

Supplier: Köttermann
These tables are available in two versions, free standing or mobile. Mobile version tables are with castors. It is suitable for writing tasks, chemical and equipment handling, hygiene and aseptic requirement areas.
In epoxy powder coated steel, with topresist® melamine or lab grade SGL worktops

  • Resistant to chemicals
  • Scratch resistance
  • Maximum loading capacity: 180 kg
  • Adjustable height



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All

Tables with TopResist® melamine worktops
Description W×D×H Pk VWR Catalog Number Quantity
Table, with melamine top-resist® worktop, free standing, with one shelf 1200×600×720/960 mm 1 KOTT505-00010
Table, with melamine top-resist® worktop, free standing, with one shelf 1500×600×720/960 mm 1 KOTT505-00011
Table, with melamine top-resist® worktop, free standing, with one shelf 900×750×720/960 mm 1 KOTT505-00012
Table, with melamine top-resist® worktop, free standing, with one shelf 1200×750×720/960 mm 1 KOTT505-00013
Table, with melamine top-resist® worktop, free standing, with one shelf 1500×750×720/960 mm 1 KOTT505-00014
Table, with melamine top-resist® worktop, free standing, with one shelf 1800×750×720/960 mm 1 KOTT505-00015
Table, with melamine top-resist® worktop, mobile, with one shelf 900×600×720/960 mm 1 KOTT505-00022
Table, with melamine top-resist® worktop, mobile, with one shelf 1200×600×720/960 mm 1 KOTT505-00023
Table, with melamine top-resist® worktop, mobile, with one shelf 1500×600×720/960 mm 1 KOTT505-00024
Table, with melamine top-resist® worktop, mobile, with one shelf 900×750×720/960 mm 1 KOTT505-00025
Table, with melamine top-resist® worktop, mobile, with one shelf 1200×750×720/960 mm 1 KOTT505-00026
Table, with melamine top-resist® worktop, mobile, with one shelf 1500×750×720/960 mm 1 KOTT505-00027
Table, with melamine top-resist® worktop, mobile, with one shelf 1800×750×720/960 mm 1 KOTT505-00028

Tables with laboratory grade SGL worktops
Description W×D×H Pk VWR Catalog Number Quantity
Table, with laboratory SGL worktop, free standing, with one shelf 900×600×720/960 mm 1 KOTT505-00016
Table, with laboratory SGL worktop, free standing, with one shelf 1200×600×720/960 mm 1 KOTT505-00017
Table, with laboratory SGL worktop, free standing, with one shelf 1500×600×720/960 mm 1 KOTT505-00018
Table, with laboratory SGL worktop, free standing, with one shelf 900×750×720/960 mm 1 KOTT505-00019
Table, with laboratory SGL worktop, free standing, with one shelf 1200×750×720/960 mm 1 KOTT505-00020
Table, with laboratory SGL worktop, free standing, with one shelf 1500×750×720/960 mm 1 KOTT505-00021
Table, with laboratory SGL worktop, mobile, with one shelf 900×600×720/960 mm 1 KOTT505-00029
Table, with laboratory SGL worktop, mobile, with one shelf 1200×600×720/960 mm 1 KOTT505-00030
Table, with laboratory SGL worktop, mobile, with one shelf 1500×600×720/960 mm 1 KOTT505-00031
Table, with laboratory SGL worktop, mobile, with one shelf 900×750×720/960 mm 1 KOTT505-00032
Table, with laboratory SGL worktop, mobile, with one shelf 1200×750×720/960 mm 1 KOTT505-00033
Table, with laboratory SGL worktop, mobile, with one shelf 1500×750×720/960 mm 1 KOTT505-00034
Table, with laboratory SGL worktop, mobile, with one shelf 1800×750×720/960 mm 1 KOTT505-00035
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty