Multi-element calibration standard in dil. nitric acid for ICP-MS
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85026.180EA 496 GBP
85026.180
Multi-element calibration standard in dil. nitric acid for ICP-MS
Standards Cationic Standards Multi-Element Standards
30 Elements : 100 mg/L As, B, Be, Fe, Se, Zn ; 10 mg/L de Ba, Bi, Cd, Co, Cr, Cu, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Rb, Sr, Te, Ti, U, V ; 1000 mg/L Ca (HNO3 2%)
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022