Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cryobiological containers for storage in racks

Supplier: VWR Collection
VWR®
The BR series is a set of cryobiological containers conceived for storage of biological samples specifically in racks. These containers are made of aluminium with a fiberglass/epoxy neck.

  • Cryotubes, cryovials and straws can be stored in liquid or vapour phase
  • Low level alarm
  • Excellent thermal performance, guarantee a long holding time and a low nitrogen consumption

All containers are equipped with racks in stainless steel and are compatible with standard cryoboxes (for 25, 81, or 100 cryotubes). Racks for 5 ml tubes or racks with drawers for goblets are also available. All racks have individual locking springs to keep the boxes or drawers in position, to prevent samples from accidentally falling out.

The containers can also be equipped with an automatic electronic level detection (DEN unit) or with a more sophisticated electronic controller (S100 or S170-10) which is capable of managing automatic filling , temperature monitoring and offer fully traceability. Please contact us for more detail.

Certifications: Compliant with EU Medical Device Directive 93/42/ECC




 

ORDER

Expand All / Collapse All

Models supplied with S170-10 electronic
Description Pk VWR Catalog Number Quantity
Cryogenic container, BR2048 M - 6 stainless steel racks with lock. rod - 5 levels for cryoboxes (25 cryovials) + S170-10 1 478-0601
Cryogenic container, BR2100 M - 6 stainless steel racks - 6 levels for cryoboxes (100 cryovials) + S170-10 1 478-0602
Cryogenic container, BR2150 M - 6 stainless steel racks - 9 levels for cryoboxes (100 cryovials) + S170-10 1 478-0603
Cryogenic container, BR2200 M - 6 stainless steel racks - 11 levels for cryoboxes (100 cryovials) + S170-10 1 478-0604

Models supplied with S170 electronic
Description Pk VWR Catalog Number Quantity
Cryogenic container, BR2048 M - 6 stainless steel racks with lock. rod - 5 levels for cryoboxes (25 cryovials) + S170 1 478-0605
Cryogenic container, BR2100 M - 6 stainless steel racks - 6 levels for cryoboxes + S170 1 478-0606
Cryogenic container, BR2150 M - 6 stainless steel racks - 9 levels for cryoboxes + S170 1 478-0607
Cryogenic container, BR2150 M - 6 stainless steel racks - 11 levels for cryoboxes + S170 1 478-0608

Models supplied with electronic - Gas phase
Description Pk VWR Catalog Number Quantity
Cryogenic container, BR2048 M gas phase - 6 stainless steel racks with lock rod - 5 levels for cryoboxes (25 cryovials) + S170 1 478-0610
Cryogenic container, BR2100 M gas phase - 6 stainless steel racks - 6 levels for cryoboxes (100 cryovials) + S170 1 478-0612
Cryogenic container, BR2150 M gas phase - 6 stainless steel racks - 9 levels for cryoboxes (100 cryovials) + S170 1 478-0614
Cryogenic container, BR2200 M gas phase - 6 stainless steel racks - 11 levels for cryoboxes (100 cryovials) + S170 1 478-0616
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Model BR2150 M BR2100 M BR2200 M BR2048 M BR2150 M BR2100 M BR2048 M BR2200 M BR2150 M BR2100 M BR2048 M
Capacity (l) 148 100 197 48,5 148 100 48,5 197 148 100 48,5
Daily evaporation (l/day) 0,65 0,27 0,65 0,27 0,65 0,27
Ø neck (mm) 215 120 215 120 215 120
Ø×H (mm) 680×920 680×748 680×1076 500×707 680×920 680×748 500×707 680×1076 680×920 680×748 500×707
No. of stages of goblets 4 2 4 2 4 2 4 2
No. of test tube racks 6
Weight, empty (kg) 56 45 62 20,5 56 45 20,5 62 56 45 20,5
Weight, full of LN₂ (kg) 174,5 126,5 221 62 174,5 126,5 62 221 174,5 126,5 62

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Ampule holders: 2 ampules Ø 12×75 mm (mini rack) 1 478-0688
Ampule holders: 5 ampules Ø12×55 mm 1 478-0687
Goblets "Marguerite" 1 478-0513
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty