Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cleaning scrubber

Supplier: Vileda Professional
This hygienic broom is designed for outlast and outperform ordinary brooms and to cover all floor and wall scrubbing needs. The abrasion resistant bristles consist of polyester PBT-filaments with excellent bend recovery and sweeping performance as well as a high resistance to a wide range of chemicals, oils, organic solvents and detergents. The strong PP plastic material withstands heavy use. Its smooth finish helps to prevent soil clinging to the surface. Non-rust staples fix the tufts and ensure long-lasting hygiene.

  • Autoclavable up to 134 °C
  • The strong PP plastic material withstands heavy use
  • The smooth finish helps to prevent soil clinging to the surface
  • Non-rust staples fix the tufts and ensure long-lasting hygiene

Recommended handle Vileda TSU 111533, universal screw-thread for German and Italian thread.




 

ORDER

Expand All / Collapse All
Description Brush head dim. Pk VWR Catalog Number Quantity
Cleaning scrubber, Superior Deck 300 mm 1 VILE145883
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping