1,3,4,6-Tetrachloro-3-6-diphenylglycouril
Supplier: G BiosciencesTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Iodo-Gen®
, NSC 4462
, 1,3,4,6-Tetrachloro-3α,6α-diphenylglycouril
Iodination reagent is designed to aid the labeling of proteins with radioactive iodine. Radioactive iodine (¹²⁵I) is routinely used by researchers to label proteins. The iodination of proteins can be performed enzymatically or chemically. It is virtually insoluble in all aqueous solutions and allows for solid phase iodination of proteins.
Formula:
C₁₆H₁₀Cl₄N₄O₂ MW: 432.088 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00027367 CAS Number: 51592-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...