Would you like to log in again?
Total immersion thermometers
Supplier: Amarell
These thermometers with mercury filling are suitable for government testing. They are designed to measure freezing points.
  • Solid stem
  • White back
  • Durable pigment
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifications: Compliant to DIN 12785.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
The main product you are looking for is no longer available for purchase. Accompanying products and services may be found elsewhere on the page.
ACCESSORIES
Expand All / Collapse All
Precision digital thermometers
Description Pk VWR Catalog Number Quantity
Protective glass tube with cork according to DIN ISO 2207 1 AMARM247320
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty