High capacity balances, IS series
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
611-8623EA 636 GBP
VWRI611-8623 VWRI611-3573 VWRI611-8624 VWRI611-8622
High capacity balances, IS series
A robust, reliable series of balances for a wide range of applications in the laboratory, but also for general industrial use. All models have a stainless steel top pan.
  • Backlit LCD with adjustable contrast
  • Rugged metal case with plastic, in-use protective cover
  • Selectable measuring units
  • Piece-counting, check-weighing, percentage weighing, animal weighing and density determination
  • Hook for below-balance weighing can be used for weighing large samples or solid density determination (software included)
  • Max. load determination (Peak hold)
Note: All these balances are now supplied without an internal battery to meet recent transport regulations. An optional external rechargeable battery (611-8684) may be purchased for portable operation.
Accessories information: Optional Tablet System (611-8683) is suitable for use with models denoted with *.
Ordering information: These balances can be equipped with optional Tablet System to expand functions and weighing actions.
Delivery information: Supplied with plastic in use protective cover.
Caution: Tablet Systems are not currently accredited for use with verified/EC-type approved balances.