Would you like to log in again?
Collagenase, from Clostridium histolyticum
Supplier: Biochrom
The treatment of tissue with collagenase effects a careful, selective reduction of the intercellular matrix and does not influence the growth ability of the cells.
For optimal results, an exactly adjusted mixture of the proteolytic enzymes is necessary. For this purpose four different types of Collagenase, CLS I, CLS II, CLS III and CLS IV, are available. The last one is normally applied with other enzymes like trypsin, elastase, or hyaluronidase. The trypsin or trypsin/EDTA generally used in cell cultures attacks the matrix only slowly, causing moreover irreversible damages of the released cells.
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Collagenase CLS I 100 mg C1-28
Collagenase CLS II 100 mg C2-28
Collagenase CLS III 100 mg C3-28
Collagenase CLS IV 100 mg C4-28
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty