Vibrating microtome, HM 650 V

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

720-2709EA 0 GBP
RAYL920130
Vibrating microtome, HM 650 V
Cut fixed and unfixed tissue samples as well as non biological specimens with the HM 650V vibrating blade microtome. Its patented vibrating system, which is completely resistant to wear, reduces artefacts by minimising the friction on the cells due to the vibrating blade. The distortion of the vibrating blade in the z-axis is stabilised via a harmonious vibration.

  • Precise vibrating system produces minimal deflection ensuring optimal specimen integrity
  • Plug-and-Play system does not require deflection adjustments
  • Sectioning and operational parameters can be stored and recalled
  • Micrometer and vibration systems enclosed in an ergonomically designed housing
  • Three modes of operation: Single, semi-manual and continuous

The cooling of the buffer solution and the specimen is guaranteed via special cooling elements. The cooling element has an insertion for a thermometer. This way, the temperature of the buffer solution can easily be controlled. The buffer tray including chuck can freely be rotated by 330°. Up to 10 stored, operator defined working settings can be called up individually and used again for sectioning. Each operator has the possibility to store their individual basic settings and use them. Four different languages are available for the software: German, English, French and Spanish. Different blades and knives can be inserted by changing one part of the blade carrier only ie. sapphire knife, injector blades or razor blades.

Ordering information: Includes cooling element with buffer solution, removable buffer tray with cooling blocks, specimen stages, tool shelf, dust cover and instruction manual.

Delivery information: Please contact your local VWR office for details of knives, blades and a full range of accessories.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR