Would you like to log in again?
Microgen® Staph ID
Supplier: MICROGEN BIOPRODUCTS LTD
Microgen® Staph ID is a sensitive and specific test for the identification of S. aureus from primary plate culture. All rapid tests use latex agglutination technology.
  • Detecting both coagulase and protein
  • Rapid and accurate identification of S. aureus in 2 minutes
  • High specifity
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Microgen® Staph ID 20 tests 20 Tests MIBIMID69CE
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Pk VWR Catalog Number Quantity
Pyr reagent required for Staph ID and Strep ID 10 ml MIBIMID61K
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty