Would you like to log in again?
ESD safety shoes, lace-up, Light, 31055/31056
Supplier: Abeba
Light
ESD saftey shoe made from smooth leather, Air Mesh inlays, reflective material and a fuel resistant PU sole.
  • Perspiration absorbing special lining with Silver Point
  • Replaceable acc Wave insole, orthopedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock absorbing system
  • Protective steel toecap (200 J)
Colour: Black or black/grey
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S1, SRA
ORDER
Expand All / Collapse All
Black
Shoe size Colour Pk VWR Catalog Number Quantity
35 Black 1 PAIR ABEB31056/35
36 Black 1 PAIR ABEB31056/36
37 Black 1 PAIR ABEB31056/37
38 Black 1 PAIR ABEB31056/38
39 Black 1 PAIR ABEB31056/39
40 Black 1 PAIR ABEB31056/40
41 Black 1 PAIR ABEB31056/41
42 Black 1 PAIR ABEB31056/42
43 Black 1 PAIR ABEB31056/43
44 Black 1 PAIR ABEB31056/44
45 Black 1 PAIR ABEB31056/45
46 Black 1 PAIR ABEB31056/46
47 Black 1 PAIR ABEB31056/47
48 Black 1 PAIR ABEB31056/48
Black/grey
Shoe size Colour Pk VWR Catalog Number Quantity
35 Black/grey 1 PAIR ABEB31055/35
36 Black/grey 1 PAIR ABEB31055/36
37 Black/grey 1 PAIR ABEB31055/37
38 Black/grey 1 PAIR ABEB31055/38
39 Black/grey 1 PAIR ABEB31055/39
40 Black/grey 1 PAIR ABEB31055/40
42 Black/grey 1 PAIR ABEB31055/42
43 Black/grey 1 PAIR ABEB31055/43
44 Black/grey 1 PAIR ABEB31055/44
45 Black/grey 1 PAIR ABEB31055/45
46 Black/grey 1 PAIR ABEB31055/46
47 Black/grey 1 PAIR ABEB31055/47
48 Black/grey 1 PAIR ABEB31055/48
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty