You Searched For: Optics Learning Activities


13  results were found

SearchResultCount:"13"

Sort Results

List View Easy View

Rate These Search Results

Supplier: DIDALAB
Description: Triangular, cast aluminium optical bench

Catalog Number: (DIDAPOD010240)
Supplier: DIDALAB
Description: Optical glass components are manufactured with high accuracy polishing and flatness tolerance.
UOM: 1 * 1 items


Catalog Number: (DIDAPOD013135)
Supplier: DIDALAB
Description: The diameter of the beam is adjustable from 1 to 12 mm.
UOM: 1 * 1 items


Supplier: DIDALAB
Description: Educational Materials, Optics, Application: Physics, Neutral density filter, d=0,9, set in an M49 ring

Supplier: DIDALAB
Description: These wide diameter lenses are used for easy classroom experiments and for aberrations underscoring.

Supplier: DIDALAB
Description: Economical, recent design of prismatic optical bench

Supplier: DIDALAB
Description: This comprehensive optical pack allows you to implement and study the basic principles of polarisation of light.

Supplier: DIDALAB
Description: Educational Materials, Optics, Application: Physics, Diode laser, blue, Class II

Supplier: DIDALAB
Description: Educational Materials, Optics, Application: Physics, Translucent screen 20×20 cm, attached to a rod with Ø 10 mm, one graduated side

Catalog Number: (DIDAPOD010615)
Supplier: DIDALAB
Description: Prism made of standard optical glass with index near to 1,52. Equilateral dispersing prisms are used for wavelength separation applications. Intended ...
UOM: 1 * 1 items


Catalog Number: (DIDAPOD013132)
Supplier: DIDALAB
Description: Extremely luminous beam, ideal for classroom experiments. Class IIIa (5 mW Power)
UOM: 1 * 1 items


Supplier: DIDALAB
Description: High quality professional gas laser, Class II

Supplier: Binhold, Paul Lehrmittel
Description: Transmission grating for spectroscopic examinations and for experiments on diffraction and interference. Suitable to resolve the sodium D lines.

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
1 - 13 of 13
no targeter for Bottom