Tubing, LDPE
Supplier: Semadeni
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SEMA2382EA 111 GBP
SEMA2382 228-2398
Tubing, LDPE
Tubing Transfer Tubing
Made of LDPE, translucent
  • Slightly flexible
  • Highly resistant to chemicals and solvents
  • Good temperature resistance