Would you like to log in again?
Beta radiation shielding rack with lid
Supplier: Deltalab
Beta safety racks, made of acrylic resin, with lid provide radiation shielding for the user against beta radiation.
  • Stackable
  • Alphanumerically identified for better sample location
  • Easy to clean
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
For tubes No. of holes W×D×H Pk VWR Catalog Number Quantity
0,5/1,5/2,0 ml 39 125×90×80 mm 1 DELTM-014.2
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty