Sample storage tubes
Supplier: Micronic
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MCRNMP22502EA 853.04 GBP
MCRNMP22502 MCRNMP32074-Z20S MCRNMP32022_U MCRNMP32074 MCRNMP32134
Sample storage tubes
Tubes Test Tubes
PP, with U or V bottom.
  • Sterile, gamma radiated
  • Snap tubes, locks tubes into rack wells to prevent sample loss from overturned racks
  • Amber tubes for light sensitive samples